Merck
Lateral resolution in NALDI MSI: back to the future.
Analytical and bioanalytical chemistry (2014-11-05)
Lukas Krasny, Oldřich Benada, Marcela Strnadova, Karel Lemr, Vladimir Havlicek
ABSTRAKT
Nanostructure-assisted laser desorption/ionization (NALDI) has been recognized as a powerful matrix-free mass spectrometry tool ideal for imaging of small molecules. In this report, the NALDI approach was compared with matrix-assisted laser desorption/ionization (MALDI) mass spectrometry in terms of sensitivity, reproducibility, and lateral resolution, which can be achieved in mass spectrometry imaging (MSI) experiments using a Nd:YAG laser. Scanning electron microscopy was used for surface topology analysis and evaluation of a putative surface-enhanced sensitivity effect, which was observed upon reduction of the laser focus. NALDI was identified as a more reproducible technique lacking MSI artifacts arising from distant tissue removal known from MALDI oversampling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tin(II) oxide, 99.99% trace metals basis
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Tin(II) oxide, ≤60 micron particle size, powder, 97%
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Yttrium aluminum oxide, nanopowder, <150 nm particle size (TEM), 99% trace metals basis
Supelco
Rhodamine 6G, analytical standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Trifluoroacetic acid, analytical standard
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%