Merck
Mycofumigation by the volatile organic compound-producing Fungus Muscodor albus induces bacterial cell death through DNA damage.
Applied and environmental microbiology (2014-12-03)
Cambria J Alpha, Manuel Campos, Christine Jacobs-Wagner, Scott A Strobel
ABSTRAKT
Muscodor albus belongs to a genus of endophytic fungi that inhibit and kill other fungi, bacteria, and insects through production of a complex mixture of volatile organic compounds (VOCs). This process of mycofumigation has found commercial application for control of human and plant pathogens, but the mechanism of the VOC toxicity is unknown. Here, the mode of action of these volatiles was investigated through a series of genetic screens and biochemical assays. A single-gene knockout screen revealed high sensitivity for Escherichia coli lacking enzymes in the pathways of DNA repair, DNA metabolic process, and response to stress when exposed to the VOCs of M. albus. Furthermore, the sensitivity of knockouts involved in the repair of specific DNA alkyl adducts suggests that the VOCs may induce alkylation. Evidence of DNA damage suggests that these adducts lead to breaks during DNA replication or transcription if not properly repaired. Additional cytotoxicity profiling indicated that during VOC exposure, E. coli became filamentous and demonstrated an increase in cellular membrane fluidity. The volatile nature of the toxic compounds produced by M. albus and their broad range of inhibition make this fungus an attractive biological agent. Understanding the antimicrobial effects and the VOC mode of action will inform the utility and safety of potential mycofumigation applications for M. albus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Phenyl-1-naphthylamine, reagent grade, 98%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, purum p.a., for separation of mineral compounds, ≥98.0% (GC)
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, 98%
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dekstroza, United States Pharmacopeia (USP) Reference Standard
Supelco
Ampicillin