Merck
  • Comprehensive analytical strategy for biomonitoring of pesticides in urine by liquid chromatography–orbitrap high resolution masss pectrometry.
Comprehensive analytical strategy for biomonitoring of pesticides in urine by liquid chromatography–orbitrap high resolution masss pectrometry.
Journal of chromatography. A (2014-12-17)
M Roca, N Leon, A Pastor, V Yusà
ABSTRAKT
In this study we propose an analytical strategy that combines a target approach for the quantitative analysis of contemporary pesticide metabolites with a comprehensive post-target screening for the identification of biomarkers of exposure to environmental contaminants in urine using liquid chromatography coupled to high-resolution mass spectrometry (LC–HRMS). The quantitative method for the target analysis of 29 urinary metabolites of organophosphate (OP) insecticides, synthetic pyrethroids, herbicides and fungicides was validated after a previous statistical optimization of the main factors governing the ion source ionization and a fragmentation study using the high energy collision dissociation (HCD) cell. The full scan accurate mass data were acquired with a resolving power of 50,000 FWHM (scan speed, 2 Hz), in both ESI+ and ESI− modes, and with and without HCD-fragmentation. The method – LOQ was lower than 3.2 μg L−1 for the majority of the analytes. For post-target screening a customized theoretical database was built, for the identification of 60 metabolites including pesticides, PAHs, phenols, and other metabolites of environmental pollutants. For identification purposes, accurate exact mass with less than 5 ppm, and diagnostic ions including isotopes and/or fragments were used. The analytical strategy was applied to 20 urine sample collected from children living in Valencia Region. Eleven target metabolites were detected with concentrations ranging from 1.18 to 131 μg L−1. Likewise, several compounds were tentatively identified in the post-target analysis belonging to the families of phthalates, phenols and parabenes. The proposed strategy is suitable for the determination of target pesticide biomarkers in urine in the framework of biomonitoring studies, and appropriate for the identification of other non-target metabolites.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Supelco
Methanol, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%