Merck
Nutritive and xenobiotic compounds in the alien algae Undaria pinnatifida from Argentine Patagonia.
Archives of environmental contamination and toxicology (2014-10-27)
M N Gil, A I Torres, M G Commendatore, C Marinho, A Arias, E Giarratano, G N Casas
ABSTRAKT
Seaweeds have been used as food since ancient times. The edible brown algae Undaria pinnatifida is native to northeast Asia; however, in 1992, the first specimens in Patagonian environments were found and, since then, have rapidly expanded. The main object of this study was to determine, for the first time in Argentina, the nutritive composition and concentrations of trace elements and hydrocarbons in these alien algae and evaluate their usefulness as food. Sexually mature U. pinnatifida samples were collected at 10-m depth in the Nuevo and San José gulfs. The first site is influenced by activities from Puerto Madryn city, and the latter place was considered as the control. Protein, dietary fiber, and mineral concentrations were similar in both gulfs and in the same order as in eastern countries. Crude protein, indigestible fiber, and calcium and magnesium concentrations were greatest in blade; lipid concentration was greatest in sporophyll; and sodium and potassium concentrations were greatest in midrib. Amino acids showed the greatest concentrations in blades, and these were greater than those reported in kelp from Japan. Cadmium (Cd), arsenic, mercury, and hydrocarbons were detected, but only Cd showed concentrations that could be a risk for consumption. In Argentina, maximum acceptable levels of these contaminants in seaweeds are not established.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
L-Threonine, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Serine, European Pharmacopoeia (EP) Reference Standard
Valine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Tyrosine, 99%
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Valine, ≥97%
Sigma-Aldrich
DL-Valine, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)