Merck
Clarithromycin-based standard triple therapy can still be effective for Helicobacter pylori eradication in some parts of the Korea.
Journal of Korean medical science (2014-09-24)
Kyu-Hyun Yoon, Sung Woon Park, Sang Wook Lee, Beom Jin Kim, Jae Gyu Kim
ABSTRAKT
We evaluated the antibiotic resistance rates and eradication rates of clarithromycin based triple therapy from 2005 to 2010 retrospectively. In addition, we investigated the mechanism of clarithromycin resistance in Helicobacter pylori strains isolated from Korean patients. Two hundred and twelve strains of H. pylori were isolated from 204 patients. H. pylori ATCC 43504 was used as the standard strain. The eradication rates of H. pylori from 2005 to 2010 were 89.3%, 82.6%, 86.3%, 87.7%, 81.8%, and 84.2%, respectively. Total eradication rate was 84.9%. DNA sequences of the 23S RNA gene in clarithromycin-resistant strains were determined. The resistance rates of H. pylori to amoxicillin, clarithromycin, metronidazole, tetracycline, ciprofloxacin, moxifloxacin, and levofloxacin were 9.0%, 8.5%, 36.3%, 0%, 14.2%, 14.2%, and 14.2%, respectively. The multidrug resistance rate of H. pylori was 16.5%. Sequence analysis of clarithromycin-resistant strains showed an A2144G mutation in 8 of 14 strains (57.1%), a T2183C mutation in 5 of 14 strains (35.7%), and double mutations of both A2144G and T2183C in 1 of 14 strains (7.1%). In the present study, triple therapy may still be an effective eradication therapy for H. pylori infections in Korea. The A2144G and T2183C mutations are mainly present in clarithromycin-resistant isolates.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Clarithromycin, United States Pharmacopeia (USP) Reference Standard
Metronidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
USP
Amoksycylina, United States Pharmacopeia (USP) Reference Standard
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Lewofloksacyna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
Metronidazole, analytical standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
USP
Tożsamość klarytromycyny, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Supelco
Levofloxacin, analytical standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)