Merck
  • Novel molecularly imprinted stir bar sorptive extraction based on an 8-electrode array for preconcentration of trace exogenous estrogens in meat.
Novel molecularly imprinted stir bar sorptive extraction based on an 8-electrode array for preconcentration of trace exogenous estrogens in meat.
Analytica chimica acta (2014-12-04)
Li Qiao, Ning Gan, Jiayu Wang, Huiju Gao, Futao Hu, Hongfei Wang, Tianhua Li
ABSTRAKT
A novel 8-electrode array as stir bar was designed for selective extraction of trace level exogenous estrogens from food samples, followed by liquid desorption and HPLC-photodiode array detection. The array consisted of 8 screen-printed electrodes and each electrode was modified with Fe3O4@meso-/macroporous TiO2 microspheres and molecularly imprinted film (m-TiMIF). The fabrication of the imprinted film coating was very simple without organic solvents and chemical grafting. Both bisphenol A (BPA) and diethylstilbestrol (DES) were employed as templates in m-TiMIF fabrication in order to enrich both targets simultaneously. Interestingly, the imprinted stir bar array showed higher extraction capacity and selectivity for BPA and DES than the non-imprinted counterpart. Meanwhile, it exhibited fast adsorption and desorption kinetics due to increased mass transport in the ultra-thin film. Importantly, the m-TiMIF coating was robust enough for at least 20 uses without obvious alteration in extraction performance. The main parameters affecting the extraction efficiency, including stir speeding, sample pH, ionic strength, extraction time, desorption solvent and time, were optimized. Under optimal experimental conditions, the limits of detection (S/N=3) of the developed method were 0.28 and 0.47 μg L(-1) for BPA and DES respectively, with enrichment factors of 32.6 and 52.8-fold. The linear ranges were 3.0-1500 μg L(-1) and 4.0-1500 μg L(-1) for BPA and DES, respectively. The m-TiMIF-coating conferred better recovery and selectivity, compared with the commercial stir bar coating. The new method was successfully applied to assess BPA and DES in pork and chicken samples with satisfactory recovery.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Titanium(IV) butoxide, purum, ≥97.0% (gravimetric)
Sigma-Aldrich
Titanium(IV) butoxide, reagent grade, 97%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Iron(III) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology