Merck
  • Toxicity studies of coumarin 6-encapsulated polystyrene nanospheres conjugated with peanut agglutinin and poly(N-vinylacetamide) as a colonoscopic imaging agent in rats.
Toxicity studies of coumarin 6-encapsulated polystyrene nanospheres conjugated with peanut agglutinin and poly(N-vinylacetamide) as a colonoscopic imaging agent in rats.
Nanomedicine : nanotechnology, biology, and medicine (2015-03-01)
Shinji Sakuma, Hironori Kumagai, Moe Shimosato, Tokio Kitamura, Kohta Mohri, Tetsuya Ikejima, Ken-Ichiro Hiwatari, Seiji Koike, Etsuo Tobita, Richard McClure, John C Gore, Wellington Pham
ABSTRAKT
We are investigating an imaging agent that detects early-stage primary colorectal cancer on the mucosal surface in real time under colonoscopic observation. The imaging agent, which is named the nanobeacon, is fluorescent nanospheres conjugated with peanut agglutinin and poly(N-vinylacetamide). Its potential use as an imaging tool for colorectal cancer has been thoroughly validated in numerous studies. Here, toxicities of the nanobeacon were assessed in rats. The nanobeacon was prepared according to the synthetic manner which is being established as the Good Manufacturing Practice-guided production. The rat study was performed in accordance with Good Laboratory Practice regulations. No nanobeacon treatment-related toxicity was observed. The no observable adverse effect levels (NOAEL) of the nanobeacon in 7-day consecutive oral administration and single intrarectal administration were estimated to be more than 1000mg/kg/day and 50mg/kg/day, respectively. We concluded that the nanobeacon could be developed as a safe diagnostic agent for colonoscopy applications. Colon cancer remains a major cause of death. Early detection can result in early treatment and thus survival. In this article, the authors tested potential systemic toxicity of coumarin 6-encapsulated polystyrene nanospheres conjugated with peanut agglutinin (PNA) and poly(N-vinylacetamide) (PNVA), which had been shown to bind specifically to colonic cancer cells and thus very promising in colonoscopic detection of cancer cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Coumarin 6, ≥99%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Coumarin 6, 98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
SAFC
L-Glutamine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
tert-Butyl methacrylate, 98%, contains 200 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M