Merck
  • Budesonide Inhibits Intracellular Infection with Non-Typeable Haemophilus influenzae Despite Its Anti-Inflammatory Effects in Respiratory Cells and Human Lung Tissue: A Role for p38 MAP Kinase.
Budesonide Inhibits Intracellular Infection with Non-Typeable Haemophilus influenzae Despite Its Anti-Inflammatory Effects in Respiratory Cells and Human Lung Tissue: A Role for p38 MAP Kinase.
Respiration; international review of thoracic diseases (2015-10-10)
Christopher Wagner, Torsten Goldmann, Kristina Rohmann, Jan Rupp, Sebastian Marwitz, Johannes Rotta Detto Loria, Stefan Limmer, Peter Zabel, Klaus Dalhoff, Daniel Drömann
ABSTRAKT
Inhaled corticosteroids (ICS) are widely used in the treatment of obstructive lung diseases. Recent data suggest a higher pneumonia risk in chronic obstructive pulmonary disease (COPD) patients treated with ICS. Since non-typeable Haemophilus influenzae (NTHi) is the most common pathogen associated with acute exacerbations of COPD, we investigated the effects of budesonide (BUD) on NTHi-induced inflammation and invasive infection. The alveolar epithelial cell line A549 and specimens of human lung tissue (HLT) were used in our experiments. Intracellular infection was determined by a lysis/culture assay of infected cells. Activated p38 mitogen-associated protein kinase (MAPK) was assessed using Western blotting and immunohistochemistry, expression of toll-like receptor 2 (TLR2) was determined by PCR, and CXCL-8 levels were measured using ELISA. Immunohistochemistry was used for detection of CXCL-8, platelet-activating factor receptor (PAF-R) and NTHi. BUD significantly reduced CXCL-8 secretion in A549 cells and lung tissue infected with NTHi. Furthermore, BUD decreased the expression of PAF-R in HLT and A549 cells. In A549 cells and HLT, BUD inhibited intracellular infection and - synergistically with NTHi - increased the expression of TLR2 (in A549 cells). TLR2 stimulation did not influence the intracellular infection of A549 cells, but p38 MAPK inhibition resulted in a significant reduction of infection. The present study adds new insights into the effects of glucocorticoids on pulmonary host defence after NTHi infection. Although the inflammatory response to infection is suppressed by BUD, interestingly, the intracellular infection is also inhibited. This effect seems to depend on the inhibition of p38 MAPK - a key enzyme in many pro-inflammatory pathways - as well as of PAF-R expression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Budesonide, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O