Merck
Upregulation of MicroRNA-126 Contributes to Endothelial Progenitor Cell Function in Deep Vein Thrombosis via Its Target PIK3R2.
Journal of cellular biochemistry (2015-02-06)
Qingyou Meng, Wenbin Wang, Xiaobin Yu, Wendong Li, Lingshang Kong, Aimin Qian, Chenglong Li, Xiaoqiang Li
ABSTRAKT
Deep vein thrombosis (DVT) is a common complication of surgery. Endothelial progenitor cells (EPCs) are recruited into resolving venous thrombi. In this report, we investigated the effects of miR-126 on EPCs function and venous thrombus resolution. We demonstrated that overexpression of miR-126 enhanced EPCs' migration and tubulogenic activity in vitro, and promoted EPCs' homing and thrombus resolving in vivo. Moreover, we identified that miR-126 directly targeted PIK3R2 and affected PI3K/Akt signaling axis. Overall, our findings demonstrated that miR-126 promoted EPCs function through suppressing PIK3R2 expression and modulation of miR-126 may represent a potential therapeutic intervention for treating DVT.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture