Merck
Differential kinetics at PK/PEPCK branch point in the cestode, Raillietina echinobothrida.
Experimental parasitology (2015-03-31)
Bidyadhar Das, Ramnath, Asim Kumar Dutta, Veena Tandon
ABSTRAKT
Pyruvate kinase (PK; EC 2.7.1.40) and phosphoenolpyruvate carboxykinase (PEPCK; EC 4.1.1.32) are essential regulatory enzymes of glucose oxidation in helminths, the PK/PEPCK branch point being the first divergent step between carbohydrate catabolism of the parasites and their hosts. Recently, PEPCK from the cestode parasite, Raillietina echinobothrida, has been purified and characterized. In order to find out the differential kinetics, if any, at PK/PEPCK branch point in the parasite, in this study, we purified and characterized the parasite PK and compared it with the parasite PEPCK. The purified PK displayed standard Michaelis-Menten kinetics with Kmapp of 77.8 µM for its substrate PEP, whereas the Kmapp was 46.9 µM for PEPCK. PEP exhibited differential kinetics at PK/PEPCK branch point of the parasite and behaved as a homotropic effector for PEPCK, but not for PK. The inhibitory constant (Ki) for genistein and daidzein (phytochemicals from Flemingia vestita) was determined and discussed. From these results, we hypothesize that PK/PEPCK branch point is a probable site for anthelmintic action.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Guanosine 5′-diphosphate sodium salt, Type I, ≥96% (HPLC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)