Merck
  • Polymethacrylate monolithic columns for hydrophilic interaction liquid chromatography prepared using a secondary surface polymerization.
Polymethacrylate monolithic columns for hydrophilic interaction liquid chromatography prepared using a secondary surface polymerization.
Journal of chromatography. A (2015-05-30)
Sinéad Currivan, Jan M Macak, Pavel Jandera
ABSTRAKT
Zwitterionic methacrylate based polymeric monolithic columns were prepared in two-step polymerizations, with reduced polymerization times. Characteristic properties such as hydrodynamic permeability, porosity, retention factors, and pore size distribution charts were used for column evaluation. A scaffold column was fabricated by polymerization of poly(lauryl methacrylate-co-tetraethyleneglycol dimethacrylate) and was used without further modification as a support for a poly(N,N-dimethyl-N-methacryloxyethyl-N-(3-sulfopropyl)ammonium betaine-co-bisphenol A glycerolate dimethacrylate) second monolith layer with zwitterionic functionality, for HILIC separations. An additional internal structure was formed by the second monolithic layer. The fabrication procedure was reproducible with RSD<5%. Field emission scanning electron microscopy has also been used to investigate column pore morphology, using a novel technique where the polymeric material is imaged directly, without coverage with a conducting film or particles. The new polar monolithic columns were used for HILIC separations of phenolic acids, flavones, nucleosides, and bases of nucleic acids, with similar efficiencies but different selectivities for zwitterionic methacrylate monolithic columns recently prepared by single step polymerization.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Propylbenzene, 98%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Adenine, ≥99%