Merck
Therapeutic efficacy of artemisinin-loaded nanoparticles in experimental visceral leishmaniasis.
Colloids and surfaces. B, Biointerfaces (2015-05-06)
Muzamil Yaqub Want, Mohammad Islamuddin, Garima Chouhan, Hani A Ozbak, Hassan A Hemeg, Anjan Kumar Dasgupta, Asoke Prasun Chattopadhyay, Farhat Afrin
ABSTRAKT
Visceral leishmaniasis (VL) is a fatal vector-borne parasitic syndrome attributable to the protozoa of the Leishmania donovani complex. The available chemotherapeutic options are not ideal due to their potential toxicity, high cost and prolonged treatment schedule. In the present study, we conjectured the use of nano drug delivery systems for plant-derived secondary metabolite; artemisinin as an alternative strategy for the treatment of experimental VL. Artemisinin-loaded poly lactic co-glycolic acid (ALPLGA) nanoparticles prepared were spherical in shape with a particle size of 220.0±15.0 nm, 29.2±2.0% drug loading and 69.0±3.3% encapsulation efficiency. ALPLGA nanoparticles administered at doses of 10 and 20mg/kg body weight showed superior antileishmanial efficacy compared with free artemisinin in BALB/c model of VL. There was a significant reduction in hepatosplenomegaly as well as in parasite load in the liver (85.0±5.4%) and spleen (82.0±2.4%) with ALPLGA nanoparticles treatment at 20mg/kg body weight compared to free artemisinin (70.3±0.6% in liver and 62.7±3.7% in spleen). In addition, ALPLGA nanoparticle treatment restored the defective host immune response in mice with established VL infection. The protection was associated with a Th1-biased immune response as evident from a positive delayed-type hypersensitivity reaction, escalated IgG2a levels, augmented lymphoproliferation and enhancement in proinflammatory cytokines (IFN-γ and IL-2) with significant suppression of Th2 cytokines (IL-10 and IL-4) after in vitro recall, compared to infected control and free artemisinin treatment. In conclusion, our results advocate superior efficacy of ALPLGA nanoparticles over free artemisinin, which was coupled with restoration of suppressed cell-mediated immunity in animal models of VL.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glycolide, ≥99%
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Artemisinin, 98%
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Vinylene carbonate, contains ≤2% BHT as stabilizer, 97%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)