Merck
Sublethal health effects in laboratory rodents from environmentally relevant exposures to oil sands contaminants.
Environmental toxicology and chemistry (2015-07-04)
Jaime Rodríguez-Estival, Michelle A North, Judit E G Smits
ABSTRAKT
Increasing activity of oil sands extraction and processing in northern Alberta is marked by ongoing controversy about the nature and extent of associated environmental impacts. Bitumen contains a mixture of toxic chemicals, including metals and residual polycyclic aromatic hydrocarbons (PAHs), whose release into the environment poses a distinct risk to the surrounding environment, plus wildlife and human health. In the present study, the authors evaluated several subclinical biomarkers of exposure and effect to mixtures of metals (Pb, Cd, and Hg) and/or PAHs (3 alkylated forms) at environmentally relevant concentrations (100-fold and 10-fold higher than the maximum dissolved concentrations found in snow, to simulate a worst-case scenario), using laboratory mice as a model for future studies of small mammals in the wild. Both metals and alkyl-PAHs exposure were associated with 1) increased relative liver, kidney, and spleen size; 2) alterations in the homeostasis of the antioxidant vitamins A and E in liver; and 3) compromised glutathione redox status in testes, with results also indicating synergistic interactions from co-exposure. The combination of morphometric and oxidative stress biomarkers provide reliable and sensitive measures of the response to contaminant exposure in a mammalian model, suggesting associated physiological costs. Based on the present experimental study, the authors propose that wild small mammals will prove to be valuable sentinel species reflecting sublethal health effects from oil sands-related contaminants. The present study's results also present a basis for the interpretation of future field data.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
9,10-Dimethylanthracene, 99%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Mercury(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Mercury(II) chloride, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
4,6-Dimethyldibenzothiophene, 97%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC