Merck
Inflammation rapidly reorganizes mouse bone marrow B cells and their environment in conjunction with early IgM responses.
Blood (2015-07-15)
Joshua M Moreau, Alexandra Berger, Megan E Nelles, Michael Mielnik, Caren Furlonger, Selena Y Cen, Rickvinder Besla, Clinton S Robbins, Christopher J Paige
ABSTRAKT
Systemic inflammation perturbs the bone marrow environment by evicting resident B cells and favoring granulopoiesis over lymphopoiesis. Despite these conditions, a subset of marrow B cell remains to become activated and produce potent acute immunoglobulin M (IgM) responses. This discrepancy is currently unresolved and a complete characterization of early perturbations in the B-cell niche has not been undertaken. Here, we show that within a few hours of challenging mice with adjuvant or cecal puncture, B cells accumulate in the bone marrow redistributed away from sinusoid vessels. This response correlates with enhanced sensitivity to CXC chemokine ligand 12 (CXCL12) but not CXCL13 or CC chemokine ligand 21. Concurrently, a number of B-cell survival and differentiation factors are elevated to produce a transiently supportive milieu. Disrupting homing dynamics with a CXC chemokine receptor 4 inhibitor reduced the formation of IgM-secreting cells. These data highlight the rapidity with which peripheral inflammation modifies the marrow compartment, and demonstrate that such modifications regulate acute IgM production within this organ. Furthermore, our study indicates that conversion to a state of emergency granulopoiesis is temporally delayed, allowing B cells opportunity to respond to antigen.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture