Merck
The interplay between hepatic stellate cells and hepatocytes in an in vitro model of NASH.
Toxicology in vitro : an international journal published in association with BIBRA (2015-07-19)
Varenka J Barbero-Becerra, Pablo J Giraudi, Norberto C Chávez-Tapia, Misael Uribe, Claudio Tiribelli, Natalia Rosso
ABSTRAKT
A complex interplay exists between hepatocytes and hepatic stellate cells (HSC) in hepatic fibrogenesis. The activation of HSCs after liver injury leads to production of extracellular matrix (ECM). Co-culture models could be useful to mimic the liver microenvironment. This study evaluates the effect of free fatty acids (FFA) on HSC cells and the interplay with hepatocytes via both soluble-mediator and cell-cell contact. The human hepatocyte cell line (HuH7) and HSC cells (LX2) were exposed to FFA for 24 h in 3 different experimental set-ups: (A) monoculture of HSC; (B) Transwell® system (effect of soluble mediators); and (C) Simultaneous Co-Culture (SCC) (cell-to-cell connections). Intracellular FFA accumulation was assessed qualitatively (microscopy) and quantitatively (flow cytometry); the activation of HSC (alpha smooth muscle actin, α-SMA) expression of ECM components were quantified by RT-PCR. FFA exposure induces intracellular fat accumulation in all the experimental set-up but the expression of α-SMA was significantly increased only in SCC. On the contrary, the expression of ECM was substantially decreased in the transwell® system. The HSC activation is independent of FFA accumulation but requires cell-to-cell interaction with hepatocyte. On the contrary, the gene regulation of ECM components seems to occur through paracrine mediators.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology