Merck
Antioxidant and antiproliferative activities of polysaccharide fractions from litchi pulp.
Food & function (2015-06-30)
Fei Huang, Ruifen Zhang, Lihong Dong, Jinxin Guo, Yuanyuan Deng, Yang Yi, Mingwei Zhang
ABSTRAKT
Three litchi polysaccharide fractions (LPFs), LP-4, LP-6 and LP-8, were obtained by fractional precipitation using 40%, 60% and 80% ethanol, respectively. The physicochemical properties, chemical antioxidant, and cellular antioxidant and antiproliferative activities of the three polysaccharide fractions were compared. LP-6 contained the highest contents of uronic acid and binding protein among the three fractions, whereas LP-8 contained the least. Amino acid composition analysis of the binding protein revealed that LP-6 contained the most acidic and aromatic amino acids. However, LP-8 contained more galactose and mannose than LP-4 and LP-6. LP-6 exhibited the highest chemical antioxidant activities, with an oxygen radical absorbance capacity of 28.14 μmol TE per g DW. LP-8 exhibited higher cellular antioxidant activity and a greater inhibitory effect on the proliferation of A549, HepG2 and MGC-803 cells at a concentration of 100-800 μg mL(-1) than LP-4 and LP-6. In summary, the different LPFs exhibited different antioxidant and antiproliferative activities with differential physicochemical properties.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
2,2′-Azobis(2-methylpropionamidine) dihydrochloride, powder or granules, 97%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water