Merck
Age-related changes in thirst, salt appetite, and arterial blood pressure in response to aldosterone-dexamethasone combination in rats.
American journal of physiology. Regulatory, integrative and comparative physiology (2015-04-03)
Robert L Thunhorst, Baojian Xue, Terry G Beltz, Alan Kim Johnson
ABSTRAKT
This work examined the effects of age on daily water and sodium ingestion and cardiovascular responses to chronic administration of the mineralocorticoid, aldosterone (ALDO) either alone or together with the glucocorticoid, dexamethasone (DEX). Young (4 mo), adult (12 mo), and aged (30 mo) male Brown Norway rats were prepared for continuous telemetry recording of blood pressure (BP) and heart rate (HR). Baseline water and sodium (i.e., 0.3 M NaCl) intake, BP, and HR were established for 10 days. Then ALDO (60 μg/day sc) was infused alone, or together with DEX (2.5 or 20 μg/day sc), for another 10 days. Compared with baseline levels, ALDO stimulated comparable increases in daily saline intake at all ages. ALDO together with the higher dose of DEX (i.e., ALDO/DEX20) increased daily saline intake more than did ALDO, but less so in aged rats. Infusion of ALDO/DEX20 increased mean arterial pressure (MAP), and decreased HR, more than did infusion of ALDO. The changes in MAP in response to both treatments depended on age. For all ages, MAP and saline intake increased simultaneously during ALDO, while MAP always increased before saline intake did during ALDO/DEX20. Contrary to our predictions, MAP did not increase more in old rats in response to either treatment. We speculate that age-related declines in cardiovascular responses to glucocorticoids contributed to the attenuated increases in sodium intake in response to glucocorticoids that were observed in older animals.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)