Merck
  • Electroacupuncture pretreatment exhibits anti-depressive effects by regulating hippocampal proteomics in rats with chronic restraint stress.
Electroacupuncture pretreatment exhibits anti-depressive effects by regulating hippocampal proteomics in rats with chronic restraint stress.
Neural regeneration research (2015-10-22)
Zhuo Guo, Ya Tu, Tian-Wei Guo, Yun-Chu Wu, Xue-Qin Yang, Lan Sun, Xin-Jing Yang, Wen-Yue Zhang, Yu Wang, Xu-Hui Zhang
ABSTRAKT
The clinical effect of electroacupuncture on depression is widely recognized. However, the signal transduction pathways and target proteins involved remain unclear. In the present study, rat models of chronic restraint stress were used to explore the mechanism by which electroacupuncture alleviates depression. Rats were randomly divided into control, model, and electroacupuncture groups. Chronic restraint stress was induced in the model and electroacupuncture groups by restraining rats for 28 days. In the electroacupuncture group, electroacupuncture pretreatment at Baihui (GV20) and Yintang (GV29) acupoints was performed daily (1 mA, 2 Hz, discontinuous wave, 20 minutes) prior to restraint for 28 days. Open field tests and body weight measurements were carried out to evaluate the depressive symptoms at specific time points. On day 28, the crossing number, rearing number, and body weights of the model group were significantly lower than those in the control group. Behavior test results indicated that rat models of depressive-like symptoms were successfully established by chronic restraint stress combined with solitary raising. On day 28, an isobaric tag for a relative and absolute quantitation-based quantitative proteomic approach was performed to identify differentially expressed proteins in hippocampal samples obtained from the model and electroacupuncture groups. The potential function of these differential proteins was predicted through the use of the Cluster of Orthologous Groups of proteins (COG) database. Twenty-seven differential proteins (uncharacteristic proteins expected) were selected from the model and electroacupuncture groups. In addition to unknown protein functions, COG are mainly concentrated in general prediction function, mechanism of signal transduction, amino acid transport and metabolism groups. This suggests that electroacupuncture improved depressive-like symptoms by regulating differential proteins, and most of these related proteins exist in nerve cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
SAFC
Glycine
SAFC
L-Threonine
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%