Merck
Studies on the Formation of Maillard and Caramelization Products from Glucosamine Incubated at 37 °C.
Journal of agricultural and food chemistry (2015-06-27)
Yuliya Hrynets, Maurice Ndagijimana, Mirko Betti
ABSTRAKT
This experiment compared the in vitro degradation of glucosamine (GlcN), N-acetylglucosamine, and glucose in the presence of NH3 incubated at 37 °C in phosphate buffer from 0.5 to 12 days. The reactions were monitored with UV-vis absorption and fluorescence emission spectroscopies, and the main products of degradation, quinoxaline derivatives of α-dicarbonyl compounds and condensation products, were determined using UHPLC-UV and Orbitrap mass spectrometry. GlcN produced two major dicarbonyl compounds, glucosone and 3-deoxyglucosone, ranging from 709 to 3245 mg/kg GlcN and from 272 to 4535 mg/kg GlcN, respectively. 3,4-Dideoxyglucosone-3-ene, glyoxal, hydroxypyruvaldehyde, methylglyoxal, and diacetyl were also detected in lower amounts compared to glucosone and 3-deoxyglucosone. Several pyrazine condensation products resulting from the reaction between dicarbonyls and GlcN were also identified. This study determined that GlcN is a significantly unstable molecule producing a high level of degradation products at 37 °C.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥95% (HPLC)
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, suitable for cell culture, BioReagent
Sigma-Aldrich
Glyoxal solution, BioReagent, ~40% in H2O (~8.8 M), Molecular Biology
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucosamine hydrochloride, ≥99% (HPLC), powder
Sigma-Aldrich
Glyoxal solution, 40 wt. % in H2O
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucosamine hydrochloride, ≥99%, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
2,3-Butanedione, 97%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-Glucosamine 6-phosphate, ≥98% (TLC)
Sigma-Aldrich
Methylglyoxal solution, ~40% in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications