Merck
  • Studies into the phenolic patterns of different tissues of pineapple (Ananas comosus [L.] Merr.) infructescence by HPLC-DAD-ESI-MS (n) and GC-MS analysis.
Studies into the phenolic patterns of different tissues of pineapple (Ananas comosus [L.] Merr.) infructescence by HPLC-DAD-ESI-MS (n) and GC-MS analysis.
Analytical and bioanalytical chemistry (2015-07-29)
Christof B Steingass, Mona P Glock, Ralf M Schweiggert, Reinhold Carle
ABSTRAKT
In a comprehensive study, more than 60 phenolic compounds were detected in methanolic extracts from different tissues of pineapple infructescence by high-performance liquid chromatography with diode array detection and electrospray ionisation multiple-stage mass spectrometry (HPLC-DAD-ESI-MS (n) ) as well as by gas chromatography-mass spectrometry (GC-MS). The analytical workflow combining both methods revealed numerous compounds assigned for the first time as pineapple constituents by their mass fragmentations. Pineapple crown tissue was characterised by depsides of p-coumaric and ferulic acid. In contrast, major phenolic compounds in pineapple pulp extracts were assigned to diverse S-p-coumaryl, S-coniferyl and S-sinapyl derivatives of glutathione, N-L-γ-glutamyl-L-cysteine and L-cysteine, which were also identified in the peel. The latter was additionally characterised by elevated concentrations of p-coumaric, ferulic and caffeic acid depsides and glycerides, respectively. Two peel-specific cyanidin hexosides were found. Elevated concentrations of isomeric N,N'-diferuloylspermidines may be a useful tool for the detection of fraudulent peel usage for pineapple juice production. Mass fragmentation pathways of characteristic pineapple constituents are proposed, and their putative biological functions are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size