Merck
DNA/HSA interaction and nuclease activity of an iron(III) amphiphilic sulfonated corrole.
Luminescence : the journal of biological and chemical luminescence (2015-03-05)
Yang Zhang, Jin-Yan Wen, Mian H R Mahmood, Xiang-Li Wang, Biao-Biao Lv, Xiao Ying, Hui Wang, Liang-Nian Ji, Hai-Yang Liu
ABSTRAKT
The DNA binding of amphiphilic iron(III) 2,17-bis(sulfonato)-5,10,15-tris(pentafluorophenyl)corrole complex (Fe-SC) was studied using spectroscopic methods and viscosity measurements. Its nuclease-like activity was examined by using pBR322 DNA as a target. The interaction of Fe-SC with human serum albumin (HSA) in vitro was also examined using multispectroscopic techniques. Experimental results revealed that Fe-SC binds to ct-DNA via an outside binding mode with a binding constant of 1.25 × 10(4) M(-1). This iron corrole also displays good activity during oxidative DNA cleavage by hydrogen peroxide or tert-butyl hydroperoxide oxidants, and high-valent (oxo)iron(V,VI) corrole intermediates may play an important role in DNA cleavage. Fe-SC exhibits much stronger binding affinity to site II than site I of HSA, indicating a selective binding tendency to HSA site II. The HSA conformational change induced by Fe-SC was confirmed by UV/Vis and CD spectroscopy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Boric acid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%