Merck
Uncovering Two Competing Switching Mechanisms for Epitaxial and Ultrathin Strontium Titanate-Based Resistive Switching Bits.
ACS nano (2015-10-09)
Markus Kubicek, Rafael Schmitt, Felix Messerschmitt, Jennifer L M Rupp
ABSTRAKT
Resistive switches based on anionic electronic conducting oxides are promising devices to replace transistor-based memories due to their excellent scalability and low power consumption. In this study, we create a model switching system by manufacturing resistive switches based on ultrathin 5 nm, epitaxial, and grain boundary-free strontium titanate thin films with subnanometer surface roughness. For our model devices, we unveil two competing nonvolatile resistive switching processes being of different polarities: one switching in clockwise and the other in counterclockwise direction. They can be activated selectively with respect to the effective switching voltage and time applied to the device. Combined analysis of both processes with electrical DC-methods and electrochemical impedance spectroscopy reveals that the first resistive switching process is filament-based and exhibits counterclockwise bipolar resistive switching. The R(OFF)/R(ON) resistance ratio of this process is extremely stable and can be tuned in the range 5-25 depending on the switching voltage and time. Excitingly, at high electric field strength a second bipolar resistive switching process was found. This process is clockwise and, therefore, reveals the opposite polarity switching direction when compared to the first one. Both processes do not obstruct each other, consequently, stable 1, 2, or even 3 crossover current-voltage (I-V) characteristics can be addressed for the memory bits. Equivalent circuit model analysis and fitting of impedance characteristics unequivocally show for the created grain boundary free switches that the oxide's defects and its carrier distribution close to the electrode interface contribute to the resistive switching mechanism. The addressability of two sets of resistive ON and OFF states in one device through electric field strength and switching time offers exciting new operation schemes for memory devices.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Nitric acid, ACS reagent, 70%