Merck
Neonatal exposure to pneumococcal phosphorylcholine modulates the development of house dust mite allergy during adult life.
Journal of immunology (Baltimore, Md. : 1950) (2015-05-10)
Preeyam S Patel, John F Kearney
ABSTRAKT
Currently, ∼20% of the global population suffers from an allergic disorder. Allergies and asthma occur at higher rates in developed and industrialized countries. It is clear that many human atopic diseases are initiated neonatally and herald more severe IgE-mediated disorders, including allergic asthma, which is driven by the priming of Th2 effector T cells. The hygiene hypothesis attempts to link the increased excessively sanitary conditions early in life to a default Th2 response and increasing allergic phenomena. Despite the substantial involvement of IgE Abs in such conditions, little attention has been paid to the effects of early microbial exposure on the B cell repertoire prior to the initiation of these diseases. In this study, we use Ab-binding assays to demonstrate that Streptococcus pneumoniae and house dust mite (HDM) bear similar phosphorylcholine (PC) epitopes. Neonatal C57BL/6 mice immunized with a PC-bearing pneumococcal vaccine expressed increased frequencies of PC-specific B cells in the lungs following sensitizing exposure to HDM as adults. Anti-PC IgM Abs in the lung decreased the interaction of HDM with pulmonary APCs and were affiliated with lowered allergy-associated cell infiltration into the lung, IgE production, development of airway hyperresponsiveness, and Th2 T cell priming. Thus, exposure of neonatal mice to PC-bearing pneumococci significantly reduced the development of HDM-induced allergic disease during adult life. Our findings demonstrate that B cells generated against conserved epitopes expressed by bacteria, encountered early in life, are also protective against the development of allergic disease during adult life.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Brefeldin A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥98% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, ~5% Water, technical grade, 95%
Latex beads, polystyrene, 3.0 μm mean particle size
Latex beads, polystyrene, 0.1 μm mean particle size
Sigma-Aldrich
Triton X-405 solution, 70% in H2O
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, BioXtra, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C