Merck
Reversion of multidrug resistance by a pH-responsive cyclodextrin-derived nanomedicine in drug resistant cancer cells.
Biomaterials (2015-07-29)
Qing Shi, Lin Zhang, Mengyu Liu, Xuelin Zhang, Xiangjun Zhang, Xiaoqiu Xu, Sha Chen, Xiaohui Li, Jianxiang Zhang
ABSTRAKT
Multidrug resistance (MDR) is one of the major problems responsible for inefficiency of cancer chemotherapy. Currently, there is still unmet demand for innovative strategies as well as effective and safe sensitizers to overcome MDR. In this study, we developed a nanosensitizer based on a pH-responsive nanoparticle (NP) derived from acetalated α-cyclodextrin (Ac-aCD). This pH-responsive NP could be effectively endocytosed by MDR cancer cells, and intracellularly transported by endolysosomal compartments. Ac-aCD NP was able to dramatically potentiate the activity of anticancer drugs including paclitaxel, docetaxel, cis-diamminedichloroplatinum, camptothecin, and doxorubicin. This sensitizing capability of Ac-aCD NP on MDR cells was resulted from the combined effects of decreased Pgp expression, attenuated Pgp ATPase activity, and the reduced intracellular ATP level. Ac-aCD NP exerted these diverse biological functions by intracellularly released α-cyclodextrin molecules, which were produced due to hydrolysis of Ac-aCD in acidic subcellular organelle. On the other hand, treatment with Ac-aCD NP showed no significant effects on the integrity of the plasma membrane, cytoskeleton, cell cycle, mitochondrial membrane potential, and apoptosis. These findings suggest that this pH-responsive NP has great potential for effective therapy of resistant cancers by combining with chemotherapeutic agents. It may also serve as a pharmacologically active nanocarrier for intracellular delivery of a plethora of antitumor drugs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
DPH, ≥98% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)