Merck
Cell Therapy With G-CSF-Mobilized Stem Cells in a Rat Osteoarthritis Model.
Cell transplantation (2014-03-29)
Ming-Wei Deng, Shih-Jung Wei, Tu-Lai Yew, Po-Hui Lee, Tzu-Yu Yang, Hen-Yi Chu, Shih-Chieh Hung
ABSTRAKT
G-CSF-mobilized peripheral blood stem cells (gm-PBSCs) offer a convenient cell source for treatment of hematopoietic and vascular disorders. Whether gm-PBSCs provide beneficial effects on skeleton diseases, such as osteoarthritis (OA), remains unknown. This study was undertaken to address the hypothesis that gm-PBSCs promote articular regeneration in OA. Here we studied the effect of single-dose intra-articular injection of gm-PBSCs from male donors delivered in hyaluronic acid (HA) on papain-induced OA in the knee joints of female Sprague-Dawley (SD) rats. Contralateral OA knee joints received single-dose HA alone and served as vehicle controls. We evaluated the histologic changes in glycosaminoglycan, type II collagen, type X collagen, modified Mankin score, and cell apoptosis rate in the articular cartilage of rat knees. We demonstrated that gm-PBSCs were mobilized to the peripheral blood via G-CSF infusion for 5 days in SD rats with increasing CD34(+) percentage up to 55-fold. We showed that gm-PBSCs inhibit progression of papain-induced OA via reducing articular surface irregularity, fibrillation, and erosion, preventing cellular necrosis and loss of chondrogenic proteins, such as glycosaminoglycan and type II collagen, at both 3 and 6 weeks after treatment. Moreover, gm-PBSCs reduced modified Mankin scores and cellular apoptosis rates compared with HA alone. Our findings demonstrate that HA plus gm-PBSCs, rather than HA alone, inhibits progression of OA in rats in vivo. Thus, intra-articular injection of gm-PBSCs is a convenient protocol for treating OA with consistent beneficial effects.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Anti-Collagen Type II (Ab-1) Mouse mAb (II-4C11), liquid, clone II-4C11, Calbiochem®
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
(±)-Ketamine hydrochloride, solid
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®