Merck
Anthrax lethal toxin-induced lung injury and treatment by activating MK2.
Journal of applied physiology (Bethesda, Md. : 1985) (2015-06-13)
Tiegang Liu, Rod R Warburton, Nicholas S Hill, Usamah S Kayyali
ABSTRAKT
Anthrax is associated with severe vascular leak, which is caused by the bacterial lethal toxin (LeTx). Pleural effusions and pulmonary edema that occur in anthrax are believed to reflect endothelial injury caused by the anthrax toxin. Since vascular leak can also be observed consistently in rats injected intravenously with LeTx, the latter might present a simple physiologically relevant animal model of acute lung injury (ALI). Such a model could be utilized in evaluating and developing better treatment for ALI or acute respiratory distress syndrome (ARDS), as other available rodent models do not consistently produce the endothelial permeability that is a major component of ARDS. The biological activity of LeTx resides in the lethal factor metalloprotease that specifically degrades MAP kinase kinases (MKKs). Recently, we showed that LeTx inactivation of p38 MAP kinase signaling via degradation of MKK3 in pulmonary vascular endothelial cells can be linked to compromise of the endothelial permeability barrier. LeTx effects were linked specifically to blocking activation of p38 substrate and MAP kinase-activated protein kinase 2 (MAPKAPK2 or MK2) and phosphorylation of the latter's substrate, heat shock protein 27 (HSP27). We have now designed a peptide that directly and specifically activates MK2, causing HSP27 phosphorylation in cells and in vivo. The MK2-activating peptide (MK2-AP) also blocks the effects of LeTx on endothelial barriers in cultured cells and reduces LeTx-induced pulmonary vascular leak in rats. Hence, MK2-AP has the therapeutic potential to counteract anthrax or pulmonary edema and vascular leak due to other causes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%