Merck
Retardation of lipid oxidation using gelatin film incorporated with longan seed extract compared with BHT.
Journal of food science and technology (2015-09-08)
Samart Sai-Ut, Soottawat Benjakul, Saroat Rawdkuen
ABSTRAKT
The aim of the present work was to apply the gelatin films with different levels of longan seed extract (LS) or butylated hydroxytoluene (BHT) on retardation of lipid oxidation in soybean oil. The films incorporated with various concentrations of aqueous LS (0, 50, 100, 300, and 500 ppm) or BHT (50, and 100 ppm) were developed. The films had transmittance percentages of 60-80 % at 570 nm and showed good light barrier properties when the concentration of LS or BHT increased. About 97 % protein solubility and 41 to 54 % water solubility were obtained for the developed films. Antioxidative activity of gelatin films incorporated with LS increased markedly with increasing storage time as indicated by the increase in DPPH radical scavenging activity (41-50 %) (P < 0.05). Films incorporated with LS or BHT showed the preventive effect on lipid oxidation of soybean oil during 30 days of storage. At the level of 500 ppm, LS provided the highest efficacy for lipid oxidation retardation as evidenced by lower conjugated diene (CD) values (P > 0.05). According to these findings, gelatin film incorporated with longan seed extract or BHT could be used as a tool to prolong the shelf-life of oily foods.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)