Merck
Combined effects of organic acids and salt depending on type of acids and pathogens in laboratory media and acidified pickle.
Journal of applied microbiology (2015-05-15)
Y-M Bae, S-Y Lee
ABSTRAKT
In this study, the effectiveness of combining each of seven types of acids with 3% salt as a treatment against pathogens was investigated in laboratory media and acidified food. When 0.5% malic, 0.5% tartaric, 0.5% citric or 0.25% phosphoric acid was combined with 3% salt, there was a higher reduction in Gram-negative bacteria (Escherichia coli O157:H7 and Salmonella Typhimurium) compared to when using acid alone. However, when 0.5% acetic, 0.5% propionic or 0.25% lactic acid was combined with 3% salt, the salt provided protection against the acid treatment. However, the antagonistic effects of acetic, propionic and lactic acid seen with Gram-negative bacteria were not observed in Listeria monocytogenes. Antagonistic effects were similarly observed when E. coli O157:H7 was treated with acetic acid and salt in food. These results show that the addition of salt increases the resistance of Gram-negative bacteria to acid treatments when using acetic, propionic and lactic acid. This study shows that antagonistic effects were observed when Gram-negative bacteria were treated with organic acids of simple structure. It may provide useful information for understanding the acid resistance mechanism of Gram-negative bacteria and developing methods for preserving acidified food.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Lactic acid, 88%, FCC
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
DL-Malic acid, 99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%