Merck
  • Development of liquid chromatography methods coupled to mass spectrometry for the analysis of substances with a wide variety of polarity in meconium.
Development of liquid chromatography methods coupled to mass spectrometry for the analysis of substances with a wide variety of polarity in meconium.
Talanta (2015-04-13)
Marie Meyer-Monath, Claudine Chatellier, Deirdre Cabooter, Florence Rouget, Isabelle Morel, Francois Lestremau
ABSTRAKT
Meconium is the first fecal excretion of newborns. This complex accumulative matrix allows assessing the exposure of the fetus to xenobiotics during the last 6 months of pregnancy. To determine the eventual effect of fetal exposure to micropollutants in this matrix, robust and sensitive analytical methods must be developed. This article describes the method development of liquid chromatography methods coupled to triple quadrupole mass spectrometry for relevant pollutants. The 28 selected target compounds had different physico-chemical properties from very polar (glyphosate) to non-polar molecules (pyrethroids). Tests were performed with three different types of columns: reversed phase, ion exchange and HILIC. As a unique method could not be determined for the simultaneous analysis of all compounds, three columns were selected and suitable chromatographic methods were optimized. Similar results were noticed for the separation of the target compounds dissolved in either meconium extract or solvent for reversed phase and ion exchange columns. However, for HILIC, the matrix had a significant influence on the peak shape and robustness of the method. Finally, the analytical methods were applied to "real" meconium samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
Phenylglyoxylic acid, 97%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
o-Cresol, ≥99%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
dimethyl phosphate, AldrichCPR