MilliporeSigma
  • Aquaporins play a role in desiccation and freeze tolerance in larvae of the goldenrod gall fly, Eurosta solidaginis.
Aquaporins play a role in desiccation and freeze tolerance in larvae of the goldenrod gall fly, Eurosta solidaginis.
The Journal of experimental biology (2008-03-18)
Benjamin N Philip, Shu-Xia Yi, Michael A Elnitsky, Richard E Lee
ABSTRACT
Survival of freezing not only requires organisms to tolerate ice formation within their body, but also depends on the rapid redistribution of water and cryoprotective compounds between intra- and extracellular compartments. Aquaporins are transmembrane proteins that serve as the major pathway through which water and small uncharged solutes (e.g. glycerol) enter and leave the cell. Consequently, we examined freeze-tolerant larvae of the goldenrod gall fly, Eurosta solidaginis, to determine whether aquaporins are present and if their presence promotes freeze tolerance of specific tissues. Immunoblotting with mammalian anti-AQP2, -AQP3 and -AQP4 revealed corresponding aquaporin homologues in E. solidaginis, whose patterns of expression varied depending on acclimation temperature and desiccation treatment. To examine the role of aquaporins in freeze tolerance, we froze fat body, midgut and salivary gland tissues in the presence and absence of mercuric chloride, an aquaporin inhibitor. Survival of fat body and midgut cells was significantly reduced when mercuric chloride was present. In contrast, survival of the salivary gland did not decrease when it was frozen with mercuric chloride. Overall, this study supports our hypothesis that naturally occurring aquaporins in E. solidaginis are regulated during desiccation and promote cell survival during freezing.
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
Ponceau S, Dye content 75 %
Sigma-Aldrich
Ponceau S, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Ponceau S solution