MilliporeSigma
  • FSH prevents porcine granulosa cells from hypoxia-induced apoptosis via activating mitophagy through the HIF-1α-PINK1-Parkin pathway.
FSH prevents porcine granulosa cells from hypoxia-induced apoptosis via activating mitophagy through the HIF-1α-PINK1-Parkin pathway.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-22)
Chengyu Li, Jiaqi Zhou, Zhaojun Liu, Jilong Zhou, Wang Yao, Jingli Tao, Ming Shen, Honglin Liu
ABSTRACT
In developing follicles, the granulosa cells (GCs) live in a hypoxic environment due to the devoid of blood supply. Upon hypoxic conditions, several types of mammalian cells have been reported to undergo apoptosis. Follicle-stimulating hormone (FSH) is known as the primary survival factor for antral follicles by preventing GCs apoptosis. Mitophagy is a type of organelle-specific autophagy that removes damaged or stressed mitochondria to maintain cellular health. This study provides the first evidence suggesting that FSH-mediated mitophagy protected porcine GCs from hypoxia-induced apoptosis. Our data showed that the GCs apoptosis caused by mitochondrial pathway upon hypoxia stress was markedly attenuated after FSH treatment, which was correlated with enhanced activation of mitophagy. Interestingly, FSH also stimulated mitochondrial biogenesis as suggested by increased expression of mitochondrial transcription factor A and nuclear respiratory factor 1 during hypoxia exposure. Notably, the protein level of hypoxia inducible factor-1α (HIF-1α) was significantly increased in hypoxic GCs following FSH treatment, accompanied by elevated mitophagic activity and dampened apoptotic signaling. Blocking HIF-1α inhibited mitophagy and restored hypoxia-induced apoptosis despite FSH treatment. Importantly, FSH promoted the expression of serine/threonine kinase PTEN induced putative kinase 1 (PINK1) and the E3 ligase Parkin during hypoxia stress through a HIF-1α dependent manner. This induced the mitophagic clearance of damaged mitochondria, hence inhibiting apoptosis by reducing cytochrome c releasing. The inhibition of HIF-1α and/or PINK1 using inhibitor or RNAi further confirmed the role of the FSH-HIF-1α-PINK1-Parkin-mitophagy axis in suppressing GC apoptosis under hypoxic conditions. These findings highlight a novel function of FSH in preserving GCs viability against hypoxic damage by activating HIF-1α-PINK1-Parkin-mediated mitophagy.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%