MilliporeSigma
  • Human Herpesvirus 6B Greatly Increases Risk of Depression by Activating Hypothalamic-Pituitary -Adrenal Axis during Latent Phase of Infection.
Human Herpesvirus 6B Greatly Increases Risk of Depression by Activating Hypothalamic-Pituitary -Adrenal Axis during Latent Phase of Infection.
iScience (2020-06-14)
Nobuyuki Kobayashi, Naomi Oka, Mayumi Takahashi, Kazuya Shimada, Azusa Ishii, Yoshitaka Tatebayashi, Masahiro Shigeta, Hiroyuki Yanagisawa, Kazuhiro Kondo
ABSTRACT
Little is known about the effect of latent-phase herpesviruses on their host. Human herpesvirus 6B (HHV-6B) is one of the most ubiquitous herpesviruses, and olfactory astrocytes are one of the most important sites of its latency. Here, we identified SITH-1, an HHV-6B latent protein specifically expressed in astrocytes. Mice induced to produce SITH-1 in their olfactory astrocytes exhibited olfactory bulb apoptosis, a hyper-activated hypothalamic-pituitary-adrenal (HPA) axis and depressive symptoms. The binding of SITH-1 to the host protein calcium-modulating ligand (CAML) to form an activated complex promoted the influx of extracellular calcium. The serum antibody titers for depressive patients with respect to this activated complex were significantly higher than for normal controls (p = 1.78 × 10-15), when the antibody positive rates were 79.8% and 24.4%, respectively, and the odds ratio was 12.2. These results suggest that, in the latent phase, HHV-6B may be involved in the onset of depression.
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Human IgG ELISA Kit, for serum, plasma