MilliporeSigma
  • High frequency acoustic nebulization for pulmonary delivery of antibiotic alternatives against Staphylococcus aureus.
High frequency acoustic nebulization for pulmonary delivery of antibiotic alternatives against Staphylococcus aureus.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2020-04-22)
Susan Marqus, Lillian Lee, Taghrid Istivan, Rachel Yoon Kyung Chang, Chaitali Dekiwadia, Hak-Kim Chan, Leslie Y Yeo
ABSTRACT
The increasing prevalence of multidrug resistant bacteria has warranted the search for new antimicrobial agents as existing antibiotics lose their potency. Among these, bacteriophage therapy, as well as the administration of specific bacteriolysis agents, i.e., lytic enzymes, have emerged as attractive alternatives. Nebulizers offer the possibility for delivering these therapeutics directly to the lung, which is particularly advantageous as a non-invasive and direct route to treat bacterial lung infections. Nevertheless, nebulizers can often result in significant degradation of the bacteriophage or protein, both structurally and functionally, due to the large stresses the aerosolization process imposes on these entities. In this work, we assess the capability of a novel low-cost and portable hybrid surface and bulk acoustic wave platform (HYDRA) to nebulize a Myoviridae bacteriophage (phage K) and lytic enzyme (lysostaphin) that specifically targets Staphylococcus aureus. Besides its efficiency in producing phage or protein-laden aerosols within the 1-5 μm respirable range for optimum delivery to the lower respiratory tract where lung infections commonly take place, we observe that the HYDRA platform-owing to the efficiency of driving the aerosolization process at relatively low powers and high frequencies (approximately 10 MHz)-does not result in appreciable denaturation of the phages or proteins, such that the loss of antimicrobial activity following nebulization is minimized. Specifically, a low (0.1 log10 (pfu/ml)) titer loss was obtained with the phages, resulting in a high viable respirable fraction of approximately 90%. Similarly, minimal loss of antimicrobial activity was obtained with lysostaphin upon nebulization wherein its minimum inhibitory concentration (0.5 μg/ml) remained unaltered as compared with the non-nebulized control. These results therefore demonstrate the potential of the HYDRA nebulization platform as a promising strategy for pulmonary administration of alternative antimicrobial agents to antibiotics for the treatment of lung diseases caused by pathogenic bacteria.
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
Sigma-Aldrich
Bacteriological agar, for molecular biology