MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Screening for toxic amyloid in yeast exemplifies the role of alternative pathway responsible for cytotoxicity.
Screening for toxic amyloid in yeast exemplifies the role of alternative pathway responsible for cytotoxicity.
PloS one (2009-03-06)
Julien Couthouis, Karine Rébora, Françoise Immel, Karine Berthelot, Michel Castroviejo, Christophe Cullin
ABSTRACT
The relationship between amyloid and toxic species is a central problem since the discovery of amyloid structures in different diseases. Despite intensive efforts in the field, the deleterious species remains unknown at the molecular level. This may reflect the lack of any structure-toxicity study based on a genetic approach. Here we show that a structure-toxicity study without any biochemical prerequisite can be successfully achieved in yeast. A PCR mutagenesis of the amyloid domain of HET-s leads to the identification of a mutant that might impair cellular viability. Cellular and biochemical analyses demonstrate that this toxic mutant forms GFP-amyloid aggregates that differ from the wild-type aggregates in their shape, size and molecular organization. The chaperone Hsp104 that helps to disassemble protein aggregates is strictly required for the cellular toxicity. Our structure-toxicity study suggests that the smallest aggregates are the most toxic, and opens a new way to analyze the relationship between structure and toxicity of amyloid species.
Sigma-Aldrich
Orange G, for NA electrophoresis
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, BioXtra, ≥97% (TLC)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, ≥97.0% (HPLC)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, BioUltra, Molecular Biology, ≥99.0% (HPLC)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, detergent for use in cell lysis
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, ≥94%
Sigma-Aldrich
Orange G, certified by the BSC, Dye content 80 %
Sigma-Aldrich
Orange G, certified by the BSC