MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Bridged androstenediol analogs as ER-beta selective SERMs.
Bioorganic & medicinal chemistry letters (2007-04-24)
Timothy A Blizzard, Candido Gude, Wanda Chan, Elizabeth T Birzin, Marina Mojena, Consuelo Tudela, Fang Chen, Kristin Knecht, Qin Su, Bryan Kraker, Mark A Holmes, Susan P Rohrer, Milton L Hammond
ABSTRACT
A series of bridged androstenediol derivatives was prepared. The bridged compounds exhibited reduced ER-beta selectivity relative to uncyclized analogs.
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
β-Estradiol, ≥98%
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Estradiol, meets USP testing specifications