MilliporeSigma
  • Beta-Pix-dynamin 2 complex promotes colorectal cancer progression by facilitating membrane dynamics.
Beta-Pix-dynamin 2 complex promotes colorectal cancer progression by facilitating membrane dynamics.
Cellular oncology (Dordrecht) (2021-09-29)
Seula Keum, Soo Jung Yang, Esther Park, TaeIn Kang, Jee-Hye Choi, Jangho Jeong, Ye Eun Hwang, Jung-Woong Kim, Dongeun Park, Sangmyung Rhee
ABSTRACT
Spatiotemporal regulation of cell membrane dynamics is a major process that promotes cancer cell invasion by acting as a driving force for cell migration. Beta-Pix (βPix), a guanine nucleotide exchange factor for Rac1, has been reported to be involved in actin-mediated cellular processes, such as cell migration, by interacting with various proteins. As yet, however, the molecular mechanisms underlying βPix-mediated cancer cell invasion remain unclear. The clinical significance of βPix was analyzed in patients with colorectal cancer (CRC) using public clinical databases. Pull-down and immunoprecipitation assays were employed to identify novel binding partners for βPix. Additionally, various cell biological assays including immunocytochemistry and time-lapse video microscopy were performed to assess the effects of βPix on CRC progression. A βPix-SH3 antibody delivery system was used to determine the effects of the βPix-Dyn2 complex in CRC cells. We found that the Src homology 3 (SH3) domain of βPix interacts with the proline-rich domain of Dynamin 2 (Dyn2), a large GTPase. The βPix-Dyn2 interaction promoted lamellipodia formation, along with plasma membrane localization of membrane-type 1 matrix metalloproteinase (MT1-MMP). Furthermore, we found that Src kinase-mediated phosphorylation of the tyrosine residue at position 442 of βPix enhanced βPix-Dyn2 complex formation. Disruption of the βPix-Dyn2 complex by βPix-SH3 antibodies targeting intracellular βPix inhibited CRC cell invasion. Our data indicate that spatiotemporal regulation of the Src-βPix-Dyn2 axis is crucial for CRC cell invasion by promoting membrane dynamics and MT1-MMP recruitment into the leading edge. The development of inhibitors that disrupt the βPix-Dyn2 complex may be a useful therapeutic strategy for CRC.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC50 = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).