MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Prediction and identification of synergistic compound combinations against pancreatic cancer cells.
Prediction and identification of synergistic compound combinations against pancreatic cancer cells.
iScience (2021-09-30)
Yasaman KalantarMotamedi, Ran Joo Choi, Siang-Boon Koh, Jo L Bramhall, Tai-Ping Fan, Andreas Bender
ABSTRACT
Resistance to current therapies is common for pancreatic cancer and hence novel treatment options are urgently needed. In this work, we developed and validated a computational method to select synergistic compound combinations based on transcriptomic profiles from both the disease and compound side, combined with a pathway scoring system, which was then validated prospectively by testing 30 compounds (and their combinations) on PANC-1 cells. Some compounds selected as single agents showed lower GI50 values than the standard of care, gemcitabine. Compounds suggested as combination agents with standard therapy gemcitabine based on the best performing scoring system showed on average 2.82-5.18 times higher synergies compared to compounds that were predicted to be active as single agents. Examples of highly synergistic in vitro validated compound pairs include gemcitabine combined with Entinostat, thioridazine, loperamide, scriptaid and Saracatinib. Hence, the computational approach presented here was able to identify synergistic compound combinations against pancreatic cancer cells.
Sigma-Aldrich
Loperamide hydrochloride
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Supelco
Digoxin, analytical standard
Sigma-Aldrich
Racecadotril, ≥98% (HPLC)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Teniposide, ≥97% (HPLC)
Sigma-Aldrich
Maprotiline hydrochloride, >99% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Phloretin, ≥99%
Sigma-Aldrich
CI−994, ≥98% (HPLC), powder
Sigma-Aldrich
Palbociclib, ≥98% (HPLC)