MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Glycan degradation promotes macroautophagy.
Proceedings of the National Academy of Sciences of the United States of America (2022-06-24)
Alice D Baudot, Victoria M-Y Wang, Josh D Leach, Jim O'Prey, Jaclyn S Long, Viola Paulus-Hock, Sergio Lilla, David M Thomson, John Greenhorn, Farah Ghaffar, Colin Nixon, Miep H Helfrich, Douglas Strathdee, Judith Pratt, Francesco Marchesi, Sara Zanivan, Kevin M Ryan
ABSTRACT
Macroautophagy promotes cellular homeostasis by delivering cytoplasmic constituents to lysosomes for degradation [Mizushima, Nat. Cell Biol. 20, 521-527 (2018)]. However, while most studies have focused on the mechanisms of protein degradation during this process, we report here that macroautophagy also depends on glycan degradation via the glycosidase, α-l-fucosidase 1 (FUCA1), which removes fucose from glycans. We show that cells lacking FUCA1 accumulate lysosomal glycans, which is associated with impaired autophagic flux. Moreover, in a mouse model of fucosidosis-a disease characterized by inactivating mutations in FUCA1 [Stepien et al., Genes (Basel) 11, E1383 (2020)]-glycan and autophagosome/autolysosome accumulation accompanies tissue destruction. Mechanistically, using lectin capture and mass spectrometry, we identified several lysosomal enzymes with altered fucosylation in FUCA1-null cells. Moreover, we show that the activity of some of these enzymes in the absence of FUCA1 can no longer be induced upon autophagy stimulation, causing retardation of autophagic flux, which involves impaired autophagosome-lysosome fusion. These findings therefore show that dysregulated glycan degradation leads to defective autophagy, which is likely a contributing factor in the etiology of fucosidosis.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks