MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Dopamine and its metabolites in cathepsin D heterozygous mice before and after MPTP administration.
Dopamine and its metabolites in cathepsin D heterozygous mice before and after MPTP administration.
Neuroscience letters (2013-02-09)
Donna Crabtree, Michaël Boyer-Guittaut, Xiaosen Ouyang, Naomi Fineberg, Jianhua Zhang
ABSTRACT
Cathepsin D (CD) is a lysosomal aspartyl protease which plays an important role in α-synuclein degradation, and neuronal survival. CD knockout mice die by post-natal day 25±1 due to intestinal necrosis. We analyzed the young adult male heterozygous mice, and found no behavior abnormalities in the heterozygous mice compared to wildtype littermates. LC3-II, p62, and α-synuclein levels are similar, while LAMP1 is higher in the striatum in CD heterozygous compared to wildtype mice. Interestingly, we found that dopamine and metabolites in the striatum and olfactory bulbs are at higher levels than wildtype littermates, while the DOPAC/DA and HVA/DA ratio remain similar between wildtype and CD heterozygous mice. In response to sub-chronic 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) administration, dopamine, DOPAC, and HVA are depleted to similar levels in the striatum in both heterozygous and wildtype mice. Dopamine synthesizing enzyme tyrosine hydroxylase, metabolic enzyme monoamine oxidase, and catechol-O-methyltransferase (COMT) levels are similar in the striatum in wildtype and heterozygous mice. These studies provide valuable information regarding how lysosomal function may contribute to neurochemical homeostasis in animal models.
Sigma-Aldrich
Cathepsin D from bovine spleen, lyophilized powder, ≥2.0 units/mg protein
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
Cathepsin D from human liver, lyophilized powder, ≥250 units/mg protein (E1%/280)