MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Structural organization and splice variants of the POLE1 gene encoding the catalytic subunit of human DNA polymerase epsilon.
Structural organization and splice variants of the POLE1 gene encoding the catalytic subunit of human DNA polymerase epsilon.
The Biochemical journal (1999-04-24)
D Huang, H Pospiech, T Kesti, J E Syväoja
ABSTRACT
The catalytic subunit of human DNA polymerase epsilon, an enzyme involved in nuclear DNA replication and repair, is encoded by the POLE1 gene. This gene is composed of 51 exons spanning at least 97 kb of genomic DNA. It was found to encode three alternative mRNA splice variants that differ in their 5'-terminal sequences and in the N-termini of the predicted proteins. A CpG island covers the promoter region for the major transcript in HeLa cells. This promoter is TATA-less and contains several putative binding sites for transcription factors typical of S-phase-up-regulated and serum-responsive promoters. Potential promoter regions were also identified for the two other alternative transcripts. Interestingly, no nuclear polyadenylation signal sequence was detected in the 3'-untranslated region, although a poly(A) tail was present. These results suggest a complicated regulatory machinery for the expression of the human POLE1 gene, including three alternative transcripts expressed from three promoters.
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2