MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Molecular cloning and high-level expression of human polymerase beta cDNA and comparison of the purified recombinant human and rat enzymes.
Molecular cloning and high-level expression of human polymerase beta cDNA and comparison of the purified recombinant human and rat enzymes.
Protein expression and purification (2000-01-29)
T A Patterson, W Little, X Cheng, S G Widen, A Kumar, W A Beard, S H Wilson
ABSTRACT
The cDNA encoding the human polymerase beta from HeLa cells was PCR amplified and cloned, and its nucleotide sequence determined. The DNA sequence is identical to the polymerase beta cDNA sequence from Tera-2 cells. Three expression strategies were employed that were designed to maximize translation initiation of the polymerase beta mRNA in Escherichia coli and all yielded a high level of human polymerase beta. The recombinant protein was purified and its properties were compared with those of the recombinant rat enzyme. The domain structure and kinetic parameters (k(cat) and K(m)) were nearly identical. A mouse IgG monoclonal antibody to the rat enzyme (mAb-10S) was approximately 10-fold less reactive with the human enzyme than with the rat enzyme as determined by ELISA.
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution