MilliporeSigma
  • Relating solubility data of parabens in liquid PEG 400 to the behaviour of PEG 4000-parabens solid dispersions.
Relating solubility data of parabens in liquid PEG 400 to the behaviour of PEG 4000-parabens solid dispersions.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2009-06-11)
Johan Unga, Farhad Tajarobi, Ove Norder, Göran Frenning, Anette Larsson
ABSTRACT
The solid state behaviour of polyethylene glycol 4000 (PEG 4000) and dispersions of a homologous series of parabens (methyl- (MP), ethyl- (EP), propyl- (PP) and butyl- (BP)) were examined and compared to the paraben solubility in liquid PEG 400. Dispersions were prepared by co-melting different amounts of paraben (5-80% (w/w)) and PEG 4000 and were studied using a combination of differential scanning calorimetry (DSC) and small and wide angle X-ray diffraction (SAXD/WAXD). Depending on the concentration of parabens in the dispersions, DSC showed melting peaks from folded and unfolded forms of PEG, a eutectic melting and melting of pure parabens. The fraction of folded PEG increased and the melting temperatures of both PEG forms decreased with increasing paraben content. In an apparent phase diagram of PP-PEG dispersions a eutectic mixture appeared above 5% PP. In addition, a melting peak corresponding to the paraben appeared for dispersion containing more than 60% PP. Similar phase diagrams were shown for the other parabens. The SAXD data and a 1D correlation function analysis revealed that MP and BP were incorporated into the amorphous domains of the lamellae of solid PEG to a higher degree than EP and PP. In addition, the lamellae thickness of PEG and the fraction of amorphous domains increased more for MP and BP compared to EP and PP. BP showed the highest solubility of the parabens followed by MP, EP and PP in both liquid and solid PEG. Furthermore, the thickness of the amorphous domains of the PEG in the different parabens-PEG dispersions could be correlated to the solubility in liquid PEG 400.
Product Description
Supelco
Ethylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Ethyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%