MilliporeSigma
  • Encapsulation and release of molecular cargos via temperature-induced vesicle-to-micelle transitions.
Encapsulation and release of molecular cargos via temperature-induced vesicle-to-micelle transitions.
Small (Weinheim an der Bergstrasse, Germany) (2010-11-13)
Qi Chen, Holger Schönherr, G Julius Vancso
ABSTRACT
Temperature-induced vesicle-to-micelle transitions of polystyrene-block-poly acrylic acid (PS(139)-b-PAA(17)) aggregates in tetrahydrofuran (THF)/H(2)O solvent mixtures are studied. For a typical system with an initial concentration of PS(139)-b-PAA(17) of 2 wt% and 50 vol% of H(2)O, the morphology of the aggregates changes from vesicles to micelles upon heating from room temperature to 45 °C. The transition temperature is found to depend on the polymer concentration as well as solvent composition. A higher polymer concentration results in a higher transition temperature. The morphological change is attributed to a change in the solvent-polymer interactions, which results in a reduction in interfacial energy. The corresponding temperature-induced morphological change is employed as a strategy for the reversible release and encapsulation of small molecules. The release of Rhodamine 110 bisamide above the transition temperature is observed as a result of the trypsin-catalyzed hydrolysis of the bisamide into Rhodamine 110. Likewise, the successful encapsulation of Rhodamine 110 below the transition temperature is proven using sodium nitrite as a chemical quencher.
Sigma-Aldrich
Rhodamine 110 chloride, Dye content ≥75 %
Sigma-Aldrich
Rhodamine 110 chloride, suitable for fluorescence, BioReagent, ≥99.0% (UV)