MilliporeSigma
  • A novel and efficient one-pot four-component tandem approach for the synthesis of pyran derivatives.
A novel and efficient one-pot four-component tandem approach for the synthesis of pyran derivatives.
Molecular diversity (2012-06-26)
Zhanyi Zhang, Yijing Xu, Jia Zheng, Bin Li, Yiqun Li
ABSTRACT
A series of new pyran derivatives are efficiently synthesized in a one-pot four-component tandem reaction via Suzuki coupling followed by a three-component reaction from readily available 4-bromobenzaldehyde, activated methylene compounds, and carbonyl compounds. Single crystal X-ray structure of the synthesized product is presented. The versatility and efficiency of the proposed methodology has been demonstrated in the synthesis of novel heterocyclic molecules.
Sigma-Aldrich
Benzaldehyde, SAJ special grade, ≥98.0%
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material