MilliporeSigma
  • [Balance of iron and copper in cadets of military school during physical exercise and next-day rest at different times of the year].
[Balance of iron and copper in cadets of military school during physical exercise and next-day rest at different times of the year].
Voenno-meditsinskii zhurnal (2013-07-03)
I P Zaĭtseva, V V Nasolodin, R A Beliakov, N P Arshinov, O N Zaĭtsev, S I Meshcheriakov
ABSTRACT
According to examination, with the help of emission spectrum analysis method, of 24 cadets-athletes of different specialization it was determined that during physical exercise in summer and in winter loss of ferrum and copper with excrements exceeded the intake if these microelements with food. The next day rise of impaction of microelements and decrease of excretion. But despite the positive balance of ferrum and copper, spend of these microelements during physical exercise wasn't compensated for the day of rest. That is why there is a possibility of microelement deficit.
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Continuous cast copper (O), BCR®, certified reference material, rod
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, rod, diam. 3.1 mm, 99.999%
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Copper, IRMM®, certified reference material, 0.1 mm foil
Copper (S, P), BCR®, certified reference material, chips
Copper (O), BCR®, certified reference material, rod
Copper, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis