MilliporeSigma
  • Aromatic hydroxylation as a potential measure of hydroxyl-radical formation in vivo. Identification of hydroxylated derivatives of salicylate in human body fluids.
Aromatic hydroxylation as a potential measure of hydroxyl-radical formation in vivo. Identification of hydroxylated derivatives of salicylate in human body fluids.
The Biochemical journal (1986-07-15)
M Grootveld, B Halliwell
ABSTRACT
Attack by .OH radicals, generated by a Fenton system, upon salicylate produces 2,3-dihydroxybenzoate and 2,5-dihydroxybenzoate as major products and catechol as a minor product. H.p.l.c. separation combined with electrochemical detection was used to identify and quantify 2,3-dihydroxybenzoate and 2,5-dihydroxybenzoate in human plasma and synovial fluid. We propose that conversion of salicylate into 2,3-dihydroxybenzoate, or of other aromatic compounds into specific hydroxylated products, may be a useful assay for .OH formation in the human body.
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure