MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Evaluation of some compression aids in tableting of roller compacted swellable core drug layer.
Evaluation of some compression aids in tableting of roller compacted swellable core drug layer.
International journal of pharmaceutics (2013-06-26)
D Golchert, E Bines, A Carmody
ABSTRACT
Swellable core technology (SCT) represents a broadly applicable oral osmotic drug delivery platform for the controlled release of drugs. SCT tablets control drug delivery by using osmosis to regulate the influx of water into the tablet's core. The tablet consists of two layers; drug layer and sweller layer, with a semi-permeable membrane coating and delivery port located in the drug layer side of the tablet. The key component of SCT formulations is polyethylene oxide (PEO), which is typically wet granulated with organic solvents to prevent rapid gel hydration observed during contact with aqueous environments. However, the use of organic solvents has their own environmental and cost considerations which make this form of processing undesirable. To overcome this issue, dry granulation can be employed. However, PEO is a very plastic material and problems may be encountered during the tableting process, when work hardening occurs upon double compression. The addition of compression aids to the drug layer will help to increase the roll force when generating ribbons - reducing fines and segregation potential - while also reducing work hardening effects which impact tablet friability. The five compression aids used in this study were microcrystalline cellulose (MCC), xylitol, di-calcium phosphate (anhydrous), lactose monohydrate and starch. The work undertaken here studies the compression properties of the drug layer blends with different levels of the five compression aids as part of the formulation. Roller compaction properties are also varied to provide granules with differing solid fractions. The results of this study indicate that addition of microcrystalline cellulose in the formulation in levels between 10% and 30% significantly improve the tablet hardness at lower tablet compression forces. Further work is required to investigate the impact on dissolution.
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Xylitol, analytical standard
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylitol
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Calcium phosphate dibasic, 98.0-105.0%
Sigma-Aldrich
Dibasic Calcium Phosphate, meets USP testing specifications