MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Post-transplantation consolidation and maintenance therapy with lenalidomide for Japanese patients with multiple myeloma.
Post-transplantation consolidation and maintenance therapy with lenalidomide for Japanese patients with multiple myeloma.
Anticancer research (2013-12-11)
Hiroyuki Takamatsu, Saori Munemoto, Ryoichi Murata, Yasushi Terasaki, Kenichi Nakajima, Shinji Nakao
ABSTRACT
Post-autologous stem cell transplantation (ASCT) consolidation and maintenance therapies in multiple myeloma (MM) have recently been the central focus of studies. However, there have been no reports of Japanese patients with MM treated with post-ASCT consolidation/maintenance therapies. We retrospectively evaluated eight Japanese patients with newly-diagnosed symptomatic MM who received ASCT after high-dose melphalan, and three to four courses of bortezomib-plus-dexamethasone and two courses of lenalidomide-plus-dexamethasone followed by maintenance lenalidomide for 6-24 months. Four patients achieved complete response (CR) after ASCT, and five patients (63%) achieved stringent CR after the consolidation and maintenance therapy; two out of these five were in molecular CR. At the median follow-up of 38 months, all patients were alive and only one patient had disease progression following post-ASCT therapy. Post-ASCT consolidation and maintenance therapy using lenalidomide may be effective in the treatment of Japanese patients with MM.
Product Description
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Thalidomide, ≥98%, powder
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)