MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A prospective study of anxiety, depression, and behavioral changes in the first year after a diagnosis of childhood acute lymphoblastic leukemia: a report from the Children's Oncology Group.
A prospective study of anxiety, depression, and behavioral changes in the first year after a diagnosis of childhood acute lymphoblastic leukemia: a report from the Children's Oncology Group.
Cancer (2014-01-30)
Regina M Myers, Lyn Balsamo, Xiaomin Lu, Meenakshi Devidas, Stephen P Hunger, William L Carroll, Naomi J Winick, Kelly W Maloney, Nina S Kadan-Lottick
ABSTRACT
The authors prospectively assessed anxiety, depression, and behavior in children with standard-risk acute lymphoblastic leukemia (SR-ALL) during the first year of therapy and identified associated risk factors. A cohort study was performed of 159 children (aged 2 years-9.99 years) with SR-ALL who were enrolled on Children's Oncology Group protocol AALL0331 at 31 sites. Parents completed the Behavior Assessment System for Children, the General Functioning Scale of the Family Assessment Device, and the Coping Health Inventory for Parents at approximately 1, 6, and 12 months after diagnosis. Overall, mean scores for anxiety, depression, aggression, and hyperactivity were similar to population norms. However, more children scored in the at-risk/clinical range for depression than the expected 15% at 1 month (21.7%; P= .022), 6 months (28.6%; P< .001), and 12 months (21.1%; P= .032). For anxiety, more children scored in the at-risk/clinical range at 1 month (25.2% vs 15%; P= .001), but then reverted to expected levels. On adjusted analysis, unhealthy family functioning was found to be predictive of anxiety (odds ratio [OR], 2.24; P= .033) and depression (OR, 2.40; P= .008). Hispanic ethnicity was associated with anxiety (OR, 3.35; P= .009). Worse physical functioning (P= .049), unmarried parents (P= .017), and less reliance on social support (P= .004) were found to be associated with depression. Emotional distress at 1 month predicted anxiety (OR, 7.11; P= .002) and depression (OR, 3.31; P= .023) at 12 months. Anxiety is a significant problem in a subpopulation of patients with SR-ALL immediately after diagnosis, whereas depression remains a significant problem for at least 1 year. Children of Hispanic ethnicity or those with unhealthy family functioning may be particularly vulnerable. These data suggest that clinicians should screen for anxiety and depression throughout the first year of therapy.
Product Description
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard