MilliporeSigma
  • Training for cervical cancer prevention programs in low-resource settings: focus on visual inspection with acetic acid and cryotherapy.
Training for cervical cancer prevention programs in low-resource settings: focus on visual inspection with acetic acid and cryotherapy.
International journal of gynaecology and obstetrics: the official organ of the International Federation of Gynaecology and Obstetrics (2005-04-13)
P D Blumenthal, M Lauterbach, J W Sellors, R Sankaranarayanan
ABSTRACT
The modern approach to cervical cancer prevention, characterized by use of cytology and multiple visits for diagnosis and treatment, has frequently proven challenging and unworkable in low-resource settings. Because of this, the Alliance for Cervical Cancer Prevention (ACCP) has made it a priority to investigate and assess alternative approaches, particularly the use of visual screening methods, such as visual inspection with acetic acid (VIA) and visual inspection with Lugol's iodine (VILI), for precancer and cancer detection and the use of cryotherapy as a precancer treatment method. As a result of ACCP experience in providing training to nurses and doctors in these techniques, it is now widely agreed that training should be competency based, combining both didactic and hands-on approaches, and should be done in a clinical setting that resembles the service-delivery conditions at the program site. This article reviews ACCP experiences and perceptions about the essentials of training in visual inspection and cryotherapy and presents some lessons learned with regard to training in these techniques in low-resource settings.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Supelco
Acetic acid, analytical standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard